missing translation for 'onlineSavingsMsg'
Learn More

DPN, Tocris Bioscience™

Catalog Number 18055052
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
1428-67-7
C15H13NO2
239.274
GHZHWDWADLAOIQ-UHFFFAOYSA-N
diarylpropionitrile, 2,3-bis 4-hydroxyphenyl propionitrile, 2,3-bis 4-hydroxyphenyl propanenitrile, 2,3-bis 4-hydroxyphenyl-propionitrile, 2,3-bis p-hydroxyphenyl propionitrile, dpn compound, benzenepropanenitrile,4-hydroxy-a-4-hydroxyphenyl, diarylpropionitril, 2,3-bis p-hydroxyphenyl-propionitrile, propionitrile, 2,3-bis p-hydroxyphenyl
102614
CHEBI:63949
2,3-bis(4-hydroxyphenyl)propanenitrile
C1=CC(=CC=C1CC(C#N)C2=CC=C(C=C2)O)O
This item is not returnable. View return policy
1428-67-7
C15H13NO2
239.274
GHZHWDWADLAOIQ-UHFFFAOYSA-N
diarylpropionitrile, 2,3-bis 4-hydroxyphenyl propionitrile, 2,3-bis 4-hydroxyphenyl propanenitrile, 2,3-bis 4-hydroxyphenyl-propionitrile, 2,3-bis p-hydroxyphenyl propionitrile, dpn compound, benzenepropanenitrile,4-hydroxy-a-4-hydroxyphenyl, diarylpropionitril, 2,3-bis p-hydroxyphenyl-propionitrile, propionitrile, 2,3-bis p-hydroxyphenyl
102614
CHEBI:63949
2,3-bis(4-hydroxyphenyl)propanenitrile
C1=CC(=CC=C1CC(C#N)C2=CC=C(C=C2)O)O

Highly potent ERβ agonist

Highly potent estrogen ERβ receptor agonist with a 70-fold selectivity over ERα (EC50 values are 0.85 and 66 nM respectively). Relaxes mesenteric arteries in vitro. Shown to regulate expression of GluR1, GluR2 and GluR3 in rat hippocampus.
Quantity 50 mg
Formula Weight 239.27
Percent Purity >99%
Chemical Name or Material DPN
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'