Läs mer
DPPA4 Antibody, mFluor Violet 450 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98786MFV450
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
DPPA4 Polyclonal antibody specifically detects DPPA4 in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifikationer
| Western Blot | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| Purified |
| Polyclonal | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human DPPA4. | |
| 55211 |