missing translation for 'onlineSavingsMsg'
Läs mer

DprE1-IN-2, MedChemExpress

Produktkod. 16493068
Encompass_Preferred
Change view
Click to view available options
Kvantitet:
5 mg
10 mg
Unit Size:
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet unitSize
16493068 10 mg 10mg
16445807 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16493068 Supplier Medchem Express Supplier No. HY100531

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress DprE1-IN-2 (compound 18) is a potent DprE1 inhibitor with an IC50 of 28 nM. DprE1-IN-2 has antituberculosis effect.

DprE1-IN-2 (compound 18) is a potent DprE1 inhibitor with an IC50 of 28 nM. DprE1-IN-2 has antituberculosis effect.

Specifications

Kemiskt namn eller material DprE1-IN-2
CAS 1615713-87-5
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C19H24N6O2
Kvantitet 10 mg
Löslighetsinformation DMSO : ≥ 33 mg/mL (89.57 mM)
LEDER O=C(C1=CN(CC2=NC=NC(N(C)C)=C2C)C3=CC(C)=CN=C31)NCCO
Molekylvikt (g/mol) 368.43
Formel vikt 368.43
Procent renhet 99.59%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.