Läs mer
DPT Hydrochloride, TRC
Produktkod. 30378112
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30378112 5 mg
1 options
Produktkod. 30378112 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD6788705MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material DPT (hydrochloride)
CAS 16382-06-2
Molekylformel C16 H24 N2 . Cl H
Kvantitet 5 mg
Synonym Dipropyltryptamine Hydrochloride, DPT HCl, N,N-Dipropyl-2-(1H-indol-3-yl)ethanamine Hydrochloride, 3-[2-(Dipropylamino)ethyl]indole Hydrochloride, N,N-Dipropyltryptamine Hydrochloride
InChI formel InChI=1S/C16H24N2.ClH/c1-3-10-18(11-4-2)12-9-14-13-17-16-8-6-5-7-15(14)16;/h5-8,13,17H,3-4,9-12H2,1-2H3;1H
LEDER Cl.CCCN(CCC)CCc1c[nH]c2ccccc12
IUPAC-namn N-[2-(1H-indol-3-yl)ethyl]-N-propylpropan-1-amine;hydrochloride
Molekylvikt (g/mol) 280.84
Formel vikt 280.1706