Läs mer
Tocris Bioscience™ DQP 1105
Produktkod. 18044452
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18044452 10 mg
18055632 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18044452 Leverantör Tocris Bioscience™ Leverantörsnummer 4491/10
GluN2C/GluN2D-selective NMDA antagonist
Noncompetitive NMDA receptor antagonist; displays over 50-fold selectivity for GluN2D- and GluN2C-containing receptors over GluN2B-, GluK2-, GluA1- and GluN2A-containing receptors (IC50 values are 2.7, 8.5, 121, 153, 198 and 206 μM, respectively). Reduces frequency of channel opening.
Please refer to IUPHAR Guide to Pharmacology for the most recent naming conventions.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 380560-89-4
Molekylformel C29H24BrN3O4
Specifikationer
Kemiskt namn eller material DQP 1105
Kvantitet 10 mg
Formel vikt 558.42
Renhet >97%
Molekylvikt (g/mol) 558.432