Learn More
Dräger™ CMS Chips: MTBE
Product Code. 12344629
Change view
Click to view available options
Measurement Range:
10 to 200
Unit Size:
Each
Product Code. Measurement Range Unit Size
12344629 10 to 200 Each
1 options
Product Code. 12344629 Supplier Dräger™ Supplier No. 6406530
  • ONLINE EXCLUSIVE PRICE
    3370.73 SEK
    2492.60 SEK / Paket med 1
    Save 878.13 SEK
    26% Av
Beräknad leverans: 11-12-2026
Suitable for 10 measurements of one gas
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Air Pressure 700 to 1100 hpa
Type MTBE CMS Chip
Humidity Range 1 to 30mg/L
Measurement Range 10 to 200
Response Time 90 to 450 sec.
Temperature Range (English) 50°F to 86°F
Temperature Range (Metric) 10°C to 30°C