Learn More
Dräger™ Short-Term Detector Tubes: Carbon Dioxide
Product Code. 10661923
Change view
Click to view available options
Measurement Range:
0.1 to 6%
0.5 to 10%
1 to 20%
100 to 3000
5 to 60%
Tube Type:
Carbon dioxide 0.1%/a2
Carbon dioxide 0.5%/a
Carbon dioxide 1%/a
Carbon dioxide 100/a
Carbon dioxide 100/a-p
Carbon dioxide 5%/A
Humidity Range:
23mg/L
30mg/L
40mg/L
50mg/L
Unit Size:
Pack of 10
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Measurement Range Tube Type Humidity Range Unit Size
10661923 0.5 to 10% Carbon dioxide 0.5%/a 50mg/L Pack of 10
10629503 100 to 3000 Carbon dioxide 100/a-p 23mg/L Pack of 10
10251161 100 to 3000 Carbon dioxide 100/a 23mg/L Pack of 10
10167340 0.1 to 6% Carbon dioxide 0.1%/a2 30mg/L Pack of 10
10113112 5 to 60% Carbon dioxide 5%/A 50mg/L Pack of 10
10615631 1 to 20% Carbon dioxide 1%/a 40mg/L Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 10661923 Supplier Dräger™ Supplier No. CH31401
  • ONLINE EXCLUSIVE PRICE
    2215.20 SEK
    1566.61 SEK / Paket med 10
    Save 648.59 SEK
    29% Av
 Restricted Item
Beräknad leverans: 03-12-2026
Detector tubes
Glass detector tubes are filled with an inert reagent carrier material (silica gel, aluminum oxide, or aluminum silicate) impregnated with an indicating reagent. Reagent produces a colorimetric indication in the presence of a particular gas, vapor, or aerosol. Concentrations can be read directly from the color change on the tube's printed scale. Tubes are hermetically sealed. Most have a two-year shelf life (when stored at room temperature). Available in packs of ten.
Note: Some tests require a pretube or comparison tube in addition to the detector tube. These tubes are included in the box of 10, thus reducing the number of detector tubes. The actual number of detector tubes is cited in the ordering charts.
Order Info
Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
Specifications
Components Measured Carbon Dioxide
For Use With (Equipment) Accuro Drager Gas Detector Pump
Humidity Range 50mg/L
Measurement Range 0.5 to 10%
No. of Strokes 1
Quantity 10
Response Time 30 sec.
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C
Tube Type Carbon dioxide 0.5%/a