Learn More
Dräger™ Short-Term Detector Tubes - Pyridine
Brand: Dräger™ 6728651
Additional Details : Weight : 0.23400kg
Description
1Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
2Safety Equipment Institute (SEI) certified.
Specifications
Pyridine 5/A | |
5ppm | |
10 | |
50°F to 86°F |
3 to 15mg/L | |
20 | |
20 min. | |
10°C to 30°C |
Safety and Handling
- Pyridine 5/A
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H318-Causes serious eye damage.
- H332-Harmful if inhaled.