Learn More
DR1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA563106
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: EVKEVLQECK TVALKRRKAS SRLENLGIPE EELLRQQQEL FAKARQQQAE LAQQEWLQMQ Q Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
DR1 Polyclonal antibody specifically detects DR1 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
DR1 | |
Polyclonal | |
Unconjugated | |
Dr1 | |
1700121L09Rik; Down-regulator of transcription 1; down-regulator of transcription 1, TBP-binding (negative cofactor 2); Dr1; Dr1l; NC2; NC2B; NC2beta; NC2-beta; negative cofactor 2; negative cofactor 2 beta; negative cofactor 2-beta; Protein Dr1; similar to down-regulator of transcription 1; TATA-binding protein-associated phosphoprotein; wu:fj15f04; zgc:152914 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1810 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q01658 | |
Dr1 | |
Recombinant protein corresponding to Human DR1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only