Learn More
Witeg™ Dropping Funnel with Pressure Equalization
Product Code. 11803881
Change view
Click to view available options
Capacity (Metric):
100 mL
500 mL
Stopcock:
ST 14/23
ST 29/32
ST-PTFE 29/32
Unit Size:
Each
Pack of 2
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Stopcock Unit Size
11803881 500 mL ST 29/32 Pack of 2
10573814 100 mL ST 14/23 -
10688115 100 mL ST-PTFE 29/32 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11803881 Supplier Witeg™ Supplier No. 1 895 007
  • ONLINE EXCLUSIVE PRICE
    3450.54 SEK
    3164.29 SEK / Paket med 2
    Save 286.25 SEK
    8% Av
Beräknad leverans: 19-01-2027
Use this funnel to slowly add air-sensitive reagents in a sealed environment. Witeg™ Dropping Funnel with Pressure Equalization facilitates the drop-wise addition of reagents.
  • Pressure equalizing.
  • Graduated.
  • Pack: 2 pieces.
Specifications
Capacity (Metric) 500 mL
Material Glass
Description Pressure equalizing
Graduated Yes
Includes Drip nozzle.
Graduations 2mL
Stopcock ST 29/32