Learn More
Fisherbrand™ Drosophila Bottles
Product Code. 10590143 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Bottom Shape:
Round
Capacity (English):
6 oz.
Material:
Polypropylene
Unit Size:
Pack of 500
Product Code. Bottom Shape Capacity (English) Material Unit Size
10590143 Round 6 oz. Polypropylene Pack of 500
1 options
Product Code. 10590143
Beräknad leverans: 22-12-2026
Add to basket
Add to basket
Wide base/narrow top design prevents fly meal from falling out when bottle is turned upside down
Drosophila bottles are available in transparent, autoclavable polypropylene and non-autoclavable polyethylene. The bottle openings are designed to accept standard paper lids, foam plugs, cotton balls and Buzz Plugs.
Round-Bottom Bottles
  • Polypropylene
  • Single use or autoclave for reuse
Square-Bottom Bottles
  • Available in polypropylene or polyethylene
  • Polypropylene provides good transparency and can be autoclaved for reuse or disposed of after single use
Specifications
Autoclavable Autoclavable
Bottom Shape Round
Capacity (English) 6 oz.
Capacity (Metric) 177 mL
Color Transparent
Diameter (English) Outer 2.36 in.
Diameter (Metric) Outer 60 mm
For Use With (Application) Straight wall design minimizes the dislodging of food, provides more efficient racking, filling and plugging
Height (English) 4 in.
Height (Metric) 103 mm
Material Polypropylene
Type Stock Bottle