Learn More
Drummond™ Original Portable Pipet-Aid™ Pipet Controller
Convenient alternative to mouth pipetting in the laboratory.
9630.50 SEK
Specifications
Autoclavable | Autoclavable |
---|---|
Type | Portable Filler/Dispenser |
For Use With (Application) | Laboratory pipetting |
For Use With (Equipment) | Glass or plastic pipets |
Includes | Pipet Controller, TC Nosepiece |
Product Code | Brand | Voltage | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Voltage | Price | Quantity & Availability | |||||
11714819
|
Drummond™
07897-32 |
220 V |
9630.50 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Easy-to-use volume control mechanism permits precise fluid intake and delivery
- Check valve protects against accidental fluid intake into the handle
- Can be used with glass or plastic pipets
- Autoclavable components
Order Info
Contact your Fisher Scientific Customer Service Representative for available replacement parts.
Certifications
CE, Euro version
Specifications
Autoclavable | |
Laboratory pipetting | |
Pipet Controller, TC Nosepiece |
Portable Filler/Dispenser | |
Glass or plastic pipets |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Drummond™ Original Portable Pipet-Aid™ Pipet Controller