Learn More
Sanplatec™ Dry Keeper Non-Auto
Product Code. 30245634
Change view
Click to view available options
Model:
C-3B
C-3BS
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model Unit Size
30245634 C-3B Each
16465314 C-3BS Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30245634 Supplier Sanplatec™ Supplier No. 0030
  • ONLINE EXCLUSIVE PRICE
    3330.00 SEK
    2067.31 SEK / Paket med 1
    Save 1262.69 SEK
    37% Av
Beräknad leverans: 12-02-2027
A non-auto type desiccator excellent for use in dehumidification applications.
  • With a convenient pan for silica gel
  • Shelf plates can be adjusted in 8 steps at 28 mm intervals to match the size of stored items
  • Horizontal design for added stability
  • Inside humidity is slightly different depending on the set room temperature and usage conditions
  • Body cabinet of PVC, frame of ABS
  • White frame, transparent sides
Specifications
Cabinet Style Front-loading
Material PVC, ABS
Product Type Desiccator
Includes Silicagel tray, 3pcs of shelves
Model C-3B