Beskrivning
DSN1 Polyclonal antibody specifically detects DSN1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifikationer
Specifikationer
| Antigen | DSN1 |
| Användningsområden | ELISA, Western Blot |
| Klassificering | Polyclonal |
| Konjugera | FITC |
| Formulering | PBS |
| Gene Alias | C20orf172, chromosome 20 open reading frame 172, dJ469A13.2, DSN1, MIND kinetochore complex component, homolog (S. cerevisiae), FLJ13346, hKNL-3, kinetochore null 3 homolog, kinetochore-associated protein DSN1 homolog, KNL3, MIS13MGC32987 |
| Värd art | Rabbit |
| Immunogen | DSN1, MIND kinetochore complex component, homolog. (Uniprot# Q9H410) |
| Reningsmetod | Immunogen affinity purified |
| Kvantitet | 0.1 mL |
| Visa mer |