Läs mer
DSRM-3716, TRC
Produktkod. 30007196
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30007196 100 mg
30003714 10 mg
30011075 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30007196 Leverantör Toronto Research Chemicals Leverantörsnummer TRCI707320100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 58142-99-7
Molekylformel C9 H6 N I
Molekylvikt (g/mol) 255.055
Synonym DSRM-3716
IUPAC-namn 5-iodoisoquinoline
LEDER Ic1cccc2cnccc12
Specifikationer
Kemiskt namn eller material 5-Iodoisoquinoline
Kvantitet 100 mg
InChI formel InChI=1S/C9H6IN/c10-9-3-1-2-7-6-11-5-4-8(7)9/h1-6H
Formel vikt 254.9545