Learn More
3M™ DT Combination Filter
For effective protection against a range of particulates, gases and vapors or combination hazards, use 3M™ Filters DT-Series. Suitable for use with certain 3M™ half and full-face tight-fitting respirators in negative-pressure mode as well as 3M™ Powered Air Respirator System PF-600 Series.
2240.00 SEK - 3740.00 SEK
Specifications
Filter Type Combination
Product Type Filters and Pre-filters
For Use With (Equipment) 3M™ Respirators, 3M™ Powered Air Respirator System
Quantity 10/Cs.
View More Specs
Products 5
Product Code Brand Model Price Quantity & Availability  
Product Code Brand Model Price Quantity & Availability  
17796973
View Documents Promotion Details
3M™
7100260627
DT-4036E
3375.00 SEK
Pack of 10
View alternative products
Save Up to 
»
Estimated Shipment: 11-06-2024
to see stock.
Add to basket
17706983
View Documents Promotion Details
3M™
7100260630
DT-4032E
2985.00 SEK
Pack of 10
View alternative products
Save Up to 
»
Estimated Shipment: 11-06-2024
to see stock.
Add to basket
17716983
View Documents Promotion Details
3M™
7100260644
DT-4035E
2405.00 SEK
Pack of 10
View alternative products
Save Up to 
»
Estimated Shipment: 11-06-2024
to see stock.
Add to basket
17726983
View Documents Promotion Details
3M™
7100260645
DT-4046E
3740.00 SEK
Pack of 10
View alternative products
Save Up to 
»
Estimated Shipment: 11-06-2024
to see stock.
Add to basket
17736983
View Documents Promotion Details
3M™
7100260646
DT-4031E
2240.00 SEK
Pack of 10
View alternative products
Save Up to 
»
Estimated Shipment: 11-06-2024
to see stock.
Add to basket
Description
Description
  • Includes specifications to provide protection against particulate hazards, gas & vapor hazards or a combination of both
  • Features a DIN40 mm threaded connection
  • Compatible with 3M™ Reusable Half Mask HF-300 Series, 3M™ Reusable Full Face Masks FF-300 Series and FF-600 Series and 3M™ Powered Air Respirator PF-600E Series
  • Please note DT-Series filters were previously known as Pro2000 Filters
Specifications
Specifications
Combination
Filters and Pre-filters
3M™ Respirators, 3M™ Powered Air Respirator System
10/Cs.