Learn More
3M™ DT Combination Filter
For effective protection against a range of particulates, gases and vapors or combination hazards, use 3M™ Filters DT-Series. Suitable for use with certain 3M™ half and full-face tight-fitting respirators in negative-pressure mode as well as 3M™ Powered Air Respirator System PF-600 Series.
2240.00 SEK - 3740.00 SEK
Specifications
Filter Type | Combination |
---|---|
Product Type | Filters and Pre-filters |
For Use With (Equipment) | 3M™ Respirators, 3M™ Powered Air Respirator System |
Quantity | 10/Cs. |
Product Code | Brand | Model | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Price | Quantity & Availability | |||||
17796973
|
3M™
7100260627 |
DT-4036E |
3375.00 SEK
Pack of 10 |
Estimated Shipment: 11-06-2024
Log in to see stock. |
|||||
17706983
|
3M™
7100260630 |
DT-4032E |
2985.00 SEK
Pack of 10 |
Estimated Shipment: 11-06-2024
Log in to see stock. |
|||||
17716983
|
3M™
7100260644 |
DT-4035E |
2405.00 SEK
Pack of 10 |
Estimated Shipment: 11-06-2024
Log in to see stock. |
|||||
17726983
|
3M™
7100260645 |
DT-4046E |
3740.00 SEK
Pack of 10 |
Estimated Shipment: 11-06-2024
Log in to see stock. |
|||||
17736983
|
3M™
7100260646 |
DT-4031E |
2240.00 SEK
Pack of 10 |
Estimated Shipment: 11-06-2024
Log in to see stock. |
|||||
Description
- Includes specifications to provide protection against particulate hazards, gas & vapor hazards or a combination of both
- Features a DIN40 mm threaded connection
- Compatible with 3M™ Reusable Half Mask HF-300 Series, 3M™ Reusable Full Face Masks FF-300 Series and FF-600 Series and 3M™ Powered Air Respirator PF-600E Series
- Please note DT-Series filters were previously known as Pro2000 Filters
Specifications
Combination | |
Filters and Pre-filters | |
3M™ Respirators, 3M™ Powered Air Respirator System | |
10/Cs. |