Läs mer
DTPA-tetra (t-Bu ester), TRC
Produktkod. 30372909
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30372909 100 mg
1 options
Produktkod. 30372909 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD700005100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material DTPA-tetra (t-Bu ester)
CAS 174267-71-1
Molekylformel C30H55N3O10
Kvantitet 100 mg
Synonym 9-(Carboxymethyl)-6,12-bis[2-(1,1-dimethylethoxy)-2-oxoethyl]-2,2-dimethyl-4-oxo-3-Oxa-6,9,12-triazatetradecan-14-oic acid 14-(1,1-dimethylethyl) ester, tetra-tBu-DTPA
InChI formel InChI=1S/C30H55N3O10/c1-27(2,3)40-23(36)18-32(19-24(37)41-28(4,5)6)15-13-31(17-22(34)35)14-16-33(20-25(38)42-29(7,8)9)21-26(39)43-30(10,11)12/h13-21H2,1-12H3,(H,34,35)
LEDER CC(C)(C)OC(=O)CN(CCN(CCN(CC(=O)OC(C)(C)C)CC(=O)OC(C)(C)C)CC(=O)O)CC(=O)OC(C)(C)C
Molekylvikt (g/mol) 617.77
Formel vikt 617.3887