Learn More
Scientific Industries SI™ Dual Microplate Foam Insert
Holds 2 plates in place while mixing. Scientific Industries SI™ Dual Microplate Foam Insert is used with the Vortex-Genie MAX.
1015.00 SEK - 1625.00 SEK
Product Code | Brand | No. of Positions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Positions | Price | Quantity & Availability | |||||
15567355
|
Scientific Industries SI™
SI-MX040 |
4 (2 inserts) |
1625.00 SEK
Each |
Estimated Shipment: 20-05-2024
Log in to see stock. |
|||||
15557355
|
Scientific Industries SI™
SI-MX030 |
2 |
1015.00 SEK
Each |
Estimated Shipment: 20-05-2024
Log in to see stock. |
|||||
Description
- Accommodates two standard footprint microplates.
- For use with Vortex-Genie™ MAX
Specifications
Foam | |
Vortex-Genie™ MAX |