Läs mer
Dulcitol Diepoxide, TRC
Produktkod. 30372761
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30372761 10 mg
1 options
Produktkod. 30372761 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD72051510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Dulcitol Diepoxide
CAS 23261-20-3
Molekylformel C6H10O4
Kvantitet 10 mg
Synonym 1,2:5,6-Dianhydrogalactitol, 1,2:5,6-Diepoxydulcitol, Dianhydrodulcitol, Dianhydrogalactitol, NSC 132313
InChI formel InChI=1S/C6H10O4/c7-5(3-1-9-3)6(8)4-2-10-4/h3-8H,1-2H2/t3-,4+,5+,6-
LEDER O[C@H]([C@H](O)[C@@H]1CO1)[C@H]2CO2
Molekylvikt (g/mol) 146.14
Formel vikt 146.0579