missing translation for 'onlineSavingsMsg'
Läs mer

Duloxetinhydroklorid 1,0 mg/ml i metanol (som fri bas), LoGiCal

Product Code. 30458135
Change view
Click to view available options
Kvantitet:
1 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30458135 1 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30458135 Supplier LoGiCal Supplier No. LGCAMP1122.0001
 Begränsad artikel
Beräknad leverans: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

LoGiCals certifierade referensmaterial som smarta, inbyggda lösningar och stabila isotopmärkta lösningar, med expertdokumentation och stöd. Från rättsmedicinska till kliniska laboratorier, välj LoGiCal för självförtroende, efterlevnad och analytisk excellens.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material Duloxetine hydrochloride
CAS 136434-34-9
Analyt- eller komponentnamn Duloxetine hydrochloride
Frakt skick Rumstemperatur
Molekylformel C18 H19 N O S . Cl H
Certifikat och efterlevnad ISO 17034
Synonym Duloxetine Hydrochloride, (S)-(+)-Duloxetine hydrochloride, (S)-(+)-N-Methyl-3-(1-naphthalenyloxy)-3-(2-thienyl)propanamine hydrochloride, (S)-Duloxetine hydrochloride, Cymbalta, Duloxetine hydrochloride, LY 248686 HCl, (gammaS)-N-methyl-gamma-(1-naphthalenyloxy)-2-thiophenepropanamine hydrochloride, (S)-N-methyl-gamma-(1-naphthalenyloxy)-2-thiophenepropanamine hydrochloride, (gammaS)-N-methyl-gamma-(1-naphthalenyloxy)-2-thiophenepropanamine hydrochloride (1:1)
InChI formel InChI=1S/C18H19NOS.ClH/c1-19-12-11-17(18-10-5-13-21-18)20-16-9-4-7-14-6-2-3-8-15(14)16;/h2-10,13,17,19H,11-12H2,1H3;1H/t17-;/m0./s1
Lösningstyp Enda lösning
LEDER Cl.CNCC[C@H](Oc1cccc2ccccc12)c3cccs3
IUPAC-namn (3S)-N-metyl-3-naftalen-1-yloxy-3-tiofen-2-ylpropan-1-amin; hydroklorid
Lösningsmedel eller matris Metanol
Industry Type Klinisk testning
Molekylvikt (g/mol) 333.88
Kvantitet 1 ml
Formel vikt 333,0954 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.