Learn More
DWK Life Sciences DURAN™ Bell Jar, with aperture in neck
Product Code. 10797481 Shop All DWK Life Sciences Products
Change view
Click to view available options
Diameter (Metric) Base:
185 mm
215 mm
315 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Base Unit Size
10797481 215 mm Each
10727881 185 mm Each
10152121 315 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10797481 Supplier DWK Life Sciences Supplier No. 244656106
  • ONLINE EXCLUSIVE PRICE
    4130.00 SEK
    3453.77 SEK / Paket med 1
    Save 676.23 SEK
    16% Av
Beräknad leverans: 03-12-2026
The flat flange bell is used for tests with a vacuum experimental plate (vacuum chamber). The bell has a ground flange and a ground joint for the insertion of measuring equipment.
By default, tests are carried out at normal pressure. The conduct of tests under vacuum (negative pressure) can have significant effects on the reaction sequence. In order to be able to observe these effects in the rough and fine vacuum range, a vacuum experiment plate and a bell jar (also called air pump bell) are required.
  • Tight closure due to standard ground joint
  • Top opening allows connection of additional equipment
  • Fired-on, highly durable white ceramic
Specifications
Diameter (Metric) Base 215 mm
Material Borosilicate Glass (3.3)
For Use With (Application) Vacuum applications; Tests in vacuum chambers
No. per Pack 1/Pk.
Autoclavable Autoclavable