Learn More
Lenz Laborglasinstrumente™ DURAN™ Borosilicate Glass Round Bottom Flasks with Conical Ground Joint
Brand:  Lenz Laborglasinstrumente™ 03002428
 View more versions of this product
Product Code. 10165350
  • 844.00 SEK / Pack of 10
Only null left
Estimated Shipment: 05-08-2025
Specifications
According to DIN EN ISO 4797 and in addition to standard
Borosilicate Glass, Duran Glass
Round
50 mL
24/29 NS Socket
51 mm
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Lenz Laborglasinstrumente™ DURAN™ Borosilicate Glass Round Bottom Flasks with Conical Ground Joint > Capacity/Socket Joint: 50mL-24/29 NS Socket