missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lenz Laborglasinstrumente™ DURAN™ Borosilicate Glass Two-neck Pear-shaped Flasks
382.66 SEK - 756.84 SEK
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
10400133
|
Lenz Laborglasinstrumente™
03221149 |
250 mL |
756.84 SEK
Each |
Estimated Shipment: 12-06-2024
Log in to see stock. |
|||||
10382561
|
Lenz Laborglasinstrumente™
03221128 |
50 mL |
382.66 SEK
Each |
Estimated Shipment: 12-06-2024
Log in to see stock. |
|||||
Specifications
Borosilicate Glass, Duran Glass | |
Pear |