Läs mer
DWK Life Sciences DURAN™ Crystallizing Dishes with Spout
Produktkod. p-4912666 Handla allt DWK Life Sciences Produkter
Kapacitet (metrisk):
2000 ml
Diameter (metrisk):
190 mm
Förpackningsstorlek:
Varje
Produktkod. 10759101
Brand: DWK Life Sciences 213115904
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
An excellent option for crystallization, heating and cooling baths and microscale experiments in the laboratory.
  • Acc. to DIN 12338
  • Transparent with labelling patch
  • Glass Type I/neutral glass as per USP, EP and JP
  • Autoclavable
  • Available in a range of capacities
TRUSTED_SUSTAINABILITY
Specifikationer
Autoklaveringsbar Autoclavable
Bottenform Flat
Kapacitet (metrisk) 2000 ml
Material Glass
Form Round
Typ Crystallizing Dish
Höjd (metrisk) 90 mm
Diameter (metrisk) 190 mm