Learn More
DWK Life Sciences DURAN™ Filter Disc Sintered Glass, Porosity 5, edge not fused
A prerequisite for successful work with glass filters is the selection of the right porosity. It is important to select a filter whose largest pore size is slightly below the grain size of the smallest particles still to be separated.
Brand:  DWK Life Sciences 251530502
 View more versions of this product
Product Code. 11629988
  • 2890.00 SEK / Pack of 10
Estimated Shipment: 01-10-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
Glass powder with a certain particle size distribution results in a porous, solid body. These bodies are used as filter material in the form of plates and candles. Made of DURAN™ (borosilicate glass 3.3). They are ideally suited for separations with strong acids or alkalis, for example, and thus offer advantages over other materials such as plastic or paper. Available in 6 different porosity classes according to ISO 4796 as well as ASTM EI28-99. Suitable for vacuum filtration with a maximum resulting pressure difference of 1 bar.
  • Edge non-melted and centered, surfaces untreated
  • Successful work through the correct choice of porosity (The largest pore size should be slightly below the grain size of the smallest particles still to be separated).
  • Available in 6 different porosity classes according to ISO 4796 as well as ASTM EI28-99.
  • Glass articles for further processing
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Borosilicate Glass 3.3
Suitable for vacuum filtration with a maximum resulting pressure difference of 1 bar
10/Pk.
30 mm