Learn More
DWK Life Sciences DURAN™ Glass Cap
The glass appendage is placed on a culture flask and thus closes it.
Brand:  DWK Life Sciences 214411805
Additional Details : Weight : 0.20000kg
 View more versions of this product
Product Code. 15221749
  • 1663.00 SEK / Pack of 10
Estimated Shipment
22-12-2020
Add to basket
Description
Description
Microbiology is an important field of biology and medicine and deals with microorganisms (bacteria, fungi, protozoa or viruses) and their habitats and conditions. For this purpose, it is essential to produce pure culture media on which the corresponding microorganisms can grow. In order to avoid the introduction of foreign substances, the bottles and flasks must be covered but not sealed gas-tight. Air exchange is essential for culture growth.
  • The Kapsenberg caps allow an exchange of air in the media bottle, creating a positive environment for the growth of microorganisms
Specifications
Specifications
Autoclavable
Closuring of Culture Media Bottles; Storage and Transport of Liquid Cultures; Working with Cell Culture Media Under Sterile Conditions in Biosafety Cabinets and Clean Hoods
10/Pk.
31 mm
Borosilicate Glass (3.3)