Learn More
DWK Life Sciences DURAN™ Glass Cap
Description
Microbiology is an important field of biology and medicine and deals with microorganisms (bacteria, fungi, protozoa or viruses) and their habitats and conditions. For this purpose, it is essential to produce pure culture media on which the corresponding microorganisms can grow. In order to avoid the introduction of foreign substances, the bottles and flasks must be covered but not sealed gas-tight. Air exchange is essential for culture growth.
- The Kapsenberg caps allow an exchange of air in the media bottle, creating a positive environment for the growth of microorganisms
Specifications
Diameter (Metric) | 31 mm |
For Use With (Application) | Closuring of culture media bottles; Storage and transport of liquid cultures; Working with cell culture media under sterile conditions in biosafety cabinets and clean hoods |
Material | Borosilicate Glass (3.3) |
Quantity | 10/Pk. |
Autoclavable | Autoclavable |