Learn More
Lenz Laborglasinstrumente™ Duran Glass Column Heads
Product Code. 10356071
Change view
Click to view available options
Length (Metric):
55mm
Unit Size:
Each
Product Code. Length (Metric) Unit Size
10356071 55mm Each
1 options
Product Code. 10356071 Supplier Lenz Laborglasinstrumente™ Supplier No. 05170144
  • ONLINE EXCLUSIVE PRICE
    12460.00 SEK
    10340.89 SEK / Paket med 1
    Save 2119.11 SEK
    17% Av
Beräknad leverans: 11-12-2026
Specifications
Includes With valve stopcock
Length (Metric) 55mm
Type Column Head
Material Duran Glass
Quantity 1Each