Learn More
Lenz Laborglasinstrumente™ Screwthread Tubes with NS Cone
Provides for secure and easy leak-tight connection of apparatus in the laboratory. Lenz Laborglasinstrumente™ Screwthread Tubes with NS Cone are manufactured from durable DURAN™ borosilicate glass.
Brand:  Lenz Laborglasinstrumente™ 01311824
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10405801
  • 205.64 SEK / Each
Estimated Shipment: 27-05-2024
to see stock.
Add to basket
Description
Description
  • Acc. to DIN 12257
  • Available in a range of socket and thread sizes
  • Minimized need for grease
  • Corrosion-free and resistant to thermal shock
  • With conical joint, featuring high taper accuracy and excellent tightness
Specifications
Specifications
1 Ea.
NS 24/29
GL 18
Duran Glass