Learn More
DWK Life Sciences DURAN™ Glass Stopper, Amber, Standard Taper Ground Joint, Octagonal flat head
Amber DURAN™ glass ground stopper.
309.00 SEK - 1670.00 SEK
Specifications
Autoclavable | No |
---|---|
Material | Boro 3.3 |
For Use With (Application) | Storage of light sensitive reagents, pharmaceuticals, and chemicals; Storage of air sensitive reagents, and chemicals; Storage of hygroscopic solids, granules and powders; Storage of pharmaceutical APIs; Sealing laboratory glassware such as flasks, reagent bottles, and mixing cylinders |
Product Code | Brand | Standard Taper Joint | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Standard Taper Joint | Price | Quantity & Availability | |||||
15271779
|
DWK Life Sciences
216270404 |
ST 12/21 |
1670.00 SEK
Pack of 10 |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
10050041
|
DWK Life Sciences
216271203 |
ST 45/40 |
309.00 SEK
Each |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
Description
Semi-hollow or solid amber DURAN™ glass stoppers can be used to close glassware items such as flasks, separatory funnels or reagent bottles with ground joint necks. The Standard Taper ground glass joint forms a tight, leak-proof seal. Use grease for heating or vacuum applications. With octagonal shaped head. Note on using DURAN™ glass stoppers with soda-lime glass bottles: If the bottle and the stopper have a temperature difference greater than 30°C, the glass stoppers can become stuck!
- USP/EP UV light protection due to external amber colouration
- Very durable and chemically resistant amber colour
- All USP Type 1 glass components and therefore suitable for storage of pharmaceutical API materials
- All USP glass components and therefore suitable for storage of corrosive and aggressive materials
- Tight closure due to standard taper ground joint
- Octagonal shaped head
Specifications
No | |
Boro 3.3 | |
Storage of light sensitive reagents, pharmaceuticals, and chemicals; Storage of air sensitive reagents, and chemicals; Storage of hygroscopic solids, granules and powders; Storage of pharmaceutical APIs; Sealing laboratory glassware such as flasks, reagent bottles, and mixing cylinders |