Learn More
Lenz Laborglasinstrumente™ Round Bottom Three-Neck Flasks
Especially made for reactions and can be used to make solutions or hold, contain and collect chemicals. Lenz Laborglasinstrumente™ Round Bottom Three-Neck Flasks are manufactured from durable DURAN™ glass.
Brand: Lenz Laborglasinstrumente™ B0424981
Additional Details : Weight : 0.10000kg
Description
- Acc. to DIN 12392
- With conical ground joints
- Parallel side necks
- With inscription label
- Corrosion-free and resistant to thermal shock
Specifications
2000 mL | |
Borosilicate Glass, Duran Glass |