Learn More
Lenz Laborglasinstrumente™ Round Bottom Three-Neck Flasks
Especially made for reactions and can be used to make solutions or hold, contain and collect chemicals. Lenz Laborglasinstrumente™ Round Bottom Three-Neck Flasks are manufactured from durable DURAN™ glass.
Brand:  Lenz Laborglasinstrumente™ B0424981
Additional Details : Weight : 0.10000kg
Product Code. 10616092
  • 1020.00 SEK / Each
Estimated Shipment: 05-07-2024
to see stock.
Add to basket
Description
Description
  • Acc. to DIN 12392
  • With conical ground joints
  • Parallel side necks
  • With inscription label
  • Corrosion-free and resistant to thermal shock
Specifications
Specifications
2000 mL
Borosilicate Glass, Duran Glass