Learn More
DWK Life Sciences DURAN™ Plastic Stopper, White Polyethylene, Octagonal flat head
Plastic stopper for flasks and bottles with glass ground joints
Brand:  DWK Life Sciences 292040209
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15202119
  • 80.45 SEK / Pack of 10
Estimated Shipment
10-11-2020
Add to basket
Description
Description
Stoppers with octagonal top made of polyethylene plastic. Conforms to DIN 12254. White with blue insert.
  • Made from polyethylene
  • Octagonal shaped head
Specifications
Specifications
No
10/Pk.
Polyethylene
Storage of Reagents, and Chemicals
ST 7/16