Learn More
DWK Life Sciences DURAN™ Plastic Stopper, White Polyethylene, Octagonal flat head
Plastic stopper for flasks and bottles with glass ground joints
Brand: DWK Life Sciences 292040406
Additional Details : Weight : 0.02000kg
Description
Stoppers with octagonal top made of polyethylene plastic. Conforms to DIN 12254. White with blue insert.
- Made from polyethylene
- Octagonal shaped head
Specifications
Storage of reagents, and chemicals | |
10/Pk. | |
No |
ST 12/21 | |
Polyethylene |