Learn More
Lenz Laborglasinstrumente™ Receiver Adapters, Straight, with Glass Olive
Provides for safe and secure connection to condensers in the laboratory. Lenz Laborglasinstrumente™ Receiver Adapters, Straight, with Glass Olive are manufactured from durable DURAN™ borosilicate glass.
Brand: Lenz Laborglasinstrumente™ 05272004
Additional Details : Weight : 0.06300kg
Description
- Acc. to DIN 12594
- With NS socket and cone
- Vacuum connection: glass olive
- High taper accuracy of the cones
- Minimized need for grease
- Corrosion-free and resistant to thermal shock
Specifications
Receiver Adapter | |
Duran | |
With NS socket, NS cone, drain tip and Glass olive vacuum connection |