Learn More
DWK Life Sciences DURAN™ Rolled Flange Bottle, without closure
The roller rim bottle is mainly used for sampling.
Brand:  DWK Life Sciences 214615409
Additional Details : Weight : 6.94000kg
 View more versions of this product
Product Code. 15211759
  • 4075.00 SEK / Pack of 10
Estimated Shipment
10-11-2020
Add to basket
Description
Description
Microbiology is an important field of biology and medicine and deals with microorganisms (bacteria, fungi, protozoa or viruses) and their habitats and conditions. While some bottles and flasks are used for the cultivation of micro-organisms, the roller rim bottles are mainly used for sampling (e.g. in the dairy industry).
  • Supplied without screw cap
Specifications
Specifications
Autoclavable
1000 mL
Boro 3.3
95 mm
Sample Collection; Storage and Transport of Liquid Cultures; Media Preparation and Cultivation; Delivery Without Closure
10/Pk.