Läs mer
DWK Life Sciences DURAN™ Skruvgängrör, GL, tung vägg, tillägg till DIN 12216
Produktkod. 10690672 Handla allt DWK Life Sciences Produkter
Standard konisk led nr:
GL 14
GL 18
GL 25
GL 45
Förpackningsstorlek:
Paket med 10
Paket med 100
Varje
Produktkod. 10690672
Brand: DWK Life Sciences 286254009
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
För användning på en mängd olika apparater med högre krav på mekanisk hållfasthet.
Thick-walled DURAN™ glass threaded tubes with GL thread are supplementary to DIN 12216 and are used for attaching to glass apparatus. Available in 5 sizes. The available accessories allow a wide range of applications.
  • With standardised GL thread according to DIN 12216
  • thick-walled
  • Safe and easy sealing and loosening of glass-glass components
  • Grease-free working possible
  • Glass articles for further processing
TRUSTED_SUSTAINABILITY
Specifikationer
Kvantitet 50/Pk.
Objektbeskrivning For attachment to glass apparatuses; Installation in glass apparatus and devices
Ledstorlek 16 mm
Standard konisk led nr GL 18
Tjocklek (metrisk) vägg 2.5 mm