Learn More
Lenz Laborglasinstrumente™ Duran™ Sprayer Heads
Product Code. 10509751
Change view
Click to view available options
Capacity (Metric):
6 mL
12 mL
100 mL
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10509751 100 mL Each
11973531 6 mL Each
11516914 12 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10509751 Supplier Lenz Laborglasinstrumente™ Supplier No. 05470004
  • ONLINE EXCLUSIVE PRICE
    550.00 SEK
    448.51 SEK / Paket med 1
    Save 101.49 SEK
    18% Av
Beräknad leverans: 03-12-2026
Specifications
Capacity (Metric) 100 mL
Material Duran Glass
Quantity 1/Pk.
Type Sprayer Head
For Use With (Equipment) Duran Complete Sprayer
Standard Taper Joint NS 19/26