Learn More
DWK Life Sciences DURAN™ Three-way Stopcock, complete with SBW-J key, T-bore
3-way tap with threaded plug.
1407.60 SEK - 2520.00 SEK
Specifications
Material | Borosilicate Glass (3.3), Polypropylene |
---|---|
Autoclavable | No |
Width (Metric) | 1.5 mm |
No. per Pack | 5/Pk. |
For Use With (Application) | Installation in glass apparatus and devices |
Product Code | Brand | Plug Bore | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Plug Bore | Price | Quantity & Availability | |||||
15834728
|
DWK Life Sciences
286418406 |
2.5 mm |
2520.00 SEK
Pack of 5 |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
15814728
|
DWK Life Sciences
286416405 |
2.0 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
T-valves are the most common form of 3-way valves. The DURAN™ three-way valve offers the following adjustment options: 1. Connection of two different attachments 2. Connection of all 3 attachments 3. Close all 3 attachments
- 3-way valve with threaded plug
- SBW-J glass threaded tap plug with T-bore
- Three-way valve offers the following setting options: 1. Connection of two different attachments 2. Connection of all 3 attachments 3. Close all 3 attachments
- Glass articles for further processing
Specifications
Borosilicate Glass (3.3), Polypropylene | |
1.5 mm | |
Installation in glass apparatus and devices |
No | |
5/Pk. |