Learn More
Lenz Laborglasinstrumente™ Weighing Bottles
For the precise and secure weighing of samples in the laboratory. Lenz Laborglasinstrumente™ Weighing Bottles are manufactured from durable DURAN™ borosilicate glass.
Brand:  Lenz Laborglasinstrumente™ 05560521
Additional Details : Weight : 0.13500kg
 View more versions of this product
Product Code. 10018830
  • 452.00 SEK / Each
Estimated Shipment
10-10-2018
Add to basket
Description
Description
  • With interchangeable NS stopper or cap
  • With inscription label
  • With fused-in size (mm) and capacity (mL)
  • Available in high- or low-forms
  • Corrosion-free and resistant to thermal shock
Specifications
Specifications
Low
6Pack
Bottle, Weighing
Interchangeable NS stopper, inscription label
NS 80/12
Bottle
Videos