Learn More
Lenz Laborglasinstrumente™ Weighing Bottles
For the precise and secure weighing of samples in the laboratory. Lenz Laborglasinstrumente™ Weighing Bottles are manufactured from durable DURAN™ borosilicate glass.
181.00 SEK - 4345.00 SEK
Products 14
Product Code Brand Height (Metric) Form Stopper Number Price Quantity & Availability  
Product Code Brand Height (Metric) Form Stopper Number Price Quantity & Availability  
10212661 View Documents Lenz Laborglasinstrumente™
05560502
25 mm Low NS 24/12 700.00 SEK
Pack of 6 View alternative products
Save Up to 
»
Estimated Shipment
07-01-2020
Add to basket
10008760 View Documents Lenz Laborglasinstrumente™
05560504
25 mm Low NS 40/12 181.00 SEK
Each View alternative products
Save Up to 
»
Estimated Shipment
07-01-2020
Add to basket
11945986 View Documents Lenz Laborglasinstrumente™
05560504
25 mm Low NS 40/12 1150.00 SEK
Pack of 6 View alternative products
Save Up to 
»
Call for latest availability.
+46 (0)31 352 32 30
Add to basket
11710964 View Documents Lenz Laborglasinstrumente™
242104104
30 mm Low NS 80/12 4345.00 SEK
Pack of 10 View alternative products
Save Up to 
»
Estimated Shipment
07-01-2020
Add to basket
10212851 View Documents Lenz Laborglasinstrumente™
05560005
40 mm Tall NS 24/12 705.00 SEK
Pack of 6 View alternative products
Save Up to 
»
Estimated Shipment
07-01-2020
Add to basket
10663432 View Documents Lenz Laborglasinstrumente™
05560015
50 mm Tall NS 40/12 965.00 SEK
Pack of 6 View alternative products
Save Up to 
»
Estimated Shipment
07-01-2020
Add to basket
10284431 View Documents Lenz Laborglasinstrumente™
05560012
50 mm Tall NS 34/12 930.00 SEK
Pack of 6 View alternative products
Save Up to 
»
Estimated Shipment
07-01-2020
Add to basket
10460993 View Documents Lenz Laborglasinstrumente™
05560008
50 mm Tall NS 29/12 770.00 SEK
Pack of 6 View alternative products
Save Up to 
»
Estimated Shipment
07-01-2020
Add to basket
10748841 View Documents Lenz Laborglasinstrumente™
05560019
65 mm Tall NS 40/12 980.00 SEK
Pack of 6 View alternative products
Save Up to 
»
Estimated Shipment
07-01-2020
Add to basket
10538531 View Documents Lenz Laborglasinstrumente™
05560022
70 mm Tall NS 34/12 1025.00 SEK
Pack of 6 View alternative products
Save Up to 
»
Estimated Shipment
07-01-2020
Add to basket
10708951 View Documents Lenz Laborglasinstrumente™
05560028
80 mm Tall NS 50/12 1295.00 SEK
Pack of 6 View alternative products
Save Up to 
»
Estimated Shipment
07-01-2020
Add to basket
10028830 View Documents Lenz Laborglasinstrumente™
05560025
80 mm Tall NS 40/12 1055.00 SEK
Pack of 6 View alternative products
Save Up to 
»
Estimated Shipment
07-01-2020
Add to basket
11740904 View Documents Lenz Laborglasinstrumente™
242112404
80 mm Tall NS 40/12 3165.00 SEK
Pack of 10 View alternative products
Save Up to 
»
Estimated Shipment
07-01-2020
Add to basket
10018830 View Documents Lenz Laborglasinstrumente™
05560521
30 mm Low NS 80/12 This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +46 31 352 32 00 for assistance
N/A View alternative products
Save Up to 
»
Description
Description
  • With interchangeable NS stopper or cap
  • With inscription label
  • With fused-in size (mm) and capacity (mL)
  • Available in high- or low-forms
  • Corrosion-free and resistant to thermal shock