Learn More
Lenz Laborglasinstrumente™ Weighing Bottles
For the precise and secure weighing of samples in the laboratory. Lenz Laborglasinstrumente™ Weighing Bottles are manufactured from durable DURAN™ borosilicate glass.
Brand: Lenz Laborglasinstrumente™ 05560504
Additional Details : Weight : 0.03900kg
Description
- With interchangeable NS stopper or cap
- With inscription label
- With fused-in size (mm) and capacity (mL)
- Available in high- or low-forms
- Corrosion-free and resistant to thermal shock
Specifications
Low | |
6Pack | |
Bottle, Weighing | |
25mm |
Interchangeable NS stopper, inscription label | |
NS 40/12 | |
Bottle |