Läs mer
MedChemExpress Durvalumab
Produktkod. 16482288
Encompass_Preferred
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
16482288 5 mg
1 options
Produktkod. 16482288 Leverantör MedChemExpress Leverantörsnummer HYP9919
MedChemExpress Durvalumab (MEDI 4736) is an humanized anti-PD-L1 monoclonal antibody. Durvalumab (MEDI4736) completely blocks the binding of PD-L1 to both PD-1 and CD80, with IC50s of 0.1 and 0.04 nM, respectively.
Specifikationer
Antigen PD-1/PD-L1
Kvantitet 5 mg
Innehåll och lagring Please store the product under the recommended conditions in the Certificate of Analysis.
Produkttyp Inhibitory Antibodies
Frakt skick Room temperature
CAS 1428935-60-7
Renhet eller kvalitetsklass 99.6%