Learn More
DUS2L Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA560405
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: RLVENDVAGI DVNMGCPKQY STKGGMGAAL LSDPDKIEKI LSTLVKGTRR PVTCKIRILP SLEDTLSLVK RIERTGIAAI AVHGRKREER PQHPVSCEVI KAIADTLSIP Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 93%.
Dihydrouridine synthase catalyzes reduction of the 5,6-double bond of a uridine residue on the displacement loop of tRNA. The resultant modified base, 5,6-dihydrouridine, appears to increase the conformational flexibility and dynamic motion of tRNA (Kato et al., 2005 [PubMed 15994936]).Specifications
DUS2L | |
Polyclonal | |
Unconjugated | |
DUS2 | |
2310016K04Rik; Dihydrouridine synthase 2; dihydrouridine synthase 2-like, SMM1 homolog; Dus2; Dus2l; hDUS2; RGD1305612; SMM1; SMM1 homolog; tRNA-dihydrouridine synthase 2-like; tRNA-dihydrouridine(20) synthase (NAD(P)(+)); tRNA-dihydrouridine(20) synthase [NAD(P)+]-like; Up-regulated in lung cancer protein 8; URLC8 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54920 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NX74 | |
DUS2 | |
Recombinant protein corresponding to Human DUS2L | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only