missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ DUSP6 (Human) Recombinant Protein
Human DUSP6 full-length ORF ( AAH03143, 1 a.a. - 381 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00001848-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MIDTLRPVPFASEMAISKTVAWLNEQLELGNERLLLMDCRPQELYESSHIESAINVAIPGIMLRRLQKGNLPVRALFTRGEDRDRFTRRCGTDTVVLYDESSSDWNENTGGESLLGLLLKKLKDEGCRAFYLEGGFSKFQAEFSLHCETNLDGSCSSSSPPLPVLGLGGLRISSDSSSDIESDLDRDPNSATDSDGSPLSNSQPSFPVEILPFLYLGCAKDSTNLDVLEEFGIKYILNVTPNLPNLFENAGEFKYKQIPISDHWSQNLSQFFPEAISFIDEARGKNCGVLVHCLAGISRSVTVTVAYLMQKLNLSMNDAYDIVKMKKSNISPNFNFMGQLLDFERTLGLSSPCDNRVPAQQLYFTTPSNQNVYQVDSLQST
Specifications
AAH03143 | |
dual specificity phosphatase 6 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DUSP6 | |
GST |
1848 | |
Wheat germ expression system | |
10 μg | |
MKP3, PYST1 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |