Läs mer
Tocris Bioscience™ DY 268
GREENER_CHOICE
Potent FXR antagonist
Brand:  Tocris Bioscience™ 5656/10
Encompass
Produktkod. 15777016
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter »
Beskrivning
Beskrivning
DY 268 is a potent FXR antagonist (IC50 = 7.5 nM). Exhibits no detectable cytotoxicity in cell-based assays.
Specifikationer
Specifikationer
1-[(3-Methoxyphenyl)methyl]-N-[4-methyl-3-(4-morpholinylsulfonyl)phenyl]-3-(4-methylphenyl)-1H-pyrazole-4-carboxamide
1609564-75-1