Läs mer
Dyclonine Hydrochloride, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Dyclonine Hydrochloride |
| CAS | 536-43-6 |
| Molekylformel | C18 H27 N O2 . Cl H |
| Kvantitet | 10 g |
| Synonym | Dyclonine Hydrochloride, 1-Propanone, 1-(4-butoxyphenyl)-3-(1-piperidinyl)-, hydrochloride (1:1), 1-Propanone, 1-(4-butoxyphenyl)-3-(1-piperidinyl)-, hydrochloride (9CI), Propiophenone, 4'-butoxy-3-piperidino-, hydrochloride (7CI,8CI), 4'-Butoxy-3-piperidinopropiophenone hydrochloride, Diclonina, Diclonine, Dyclocaine hydrochloride, Dyclone, Dyclonine, Dyclonine hydrochloride, Dyclothane, Tanaclone |
| InChI formel | InChI=1S/C18H27NO2.ClH/c1-2-3-15-21-17-9-7-16(8-10-17)18(20)11-14-19-12-5-4-6-13-19;/h7-10H,2-6,11-15H2,1H3;1H |
| LEDER | Cl.CCCCOc1ccc(cc1)C(=O)CCN2CCCCC2 |
| IUPAC-namn | 1-(4-butoxyphenyl)-3-piperidin-1-ylpropan-1-one;hydrochloride |
| Molekylvikt (g/mol) | 325.87 |
| Formel vikt | 325.1809 |