missing translation for 'onlineSavingsMsg'
Learn More

DYDC1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 26327-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16824835

  • 1236.20 SEK / 20µL
Estimated Shipment: 07-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a member of a family of proteins that contains a DPY30 domain. The encoded protein is involved in acrosome formation during spermatid development. This gene locus overlaps with a closely related gene on the opposite strand. Alternative splicing results in multiple transcript variants.
Specifications

Specifications

DYDC1
0.53 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8WWB3, Q9D9T0
DPY30 domain containing 1, DPY30D1, DYDC1, RSD 9
Rabbit
IgG
20 μL
-20°C
Primary
143241, 69496
Western Blot
Unconjugated
DYDC1
Liquid
Dydc1
DYDC1 Fusion Protein Ag23693
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Mouse, Human
Videos
SDS
Documents
Certificates
Special Offers

Special Offers