Learn More
Applied Biosystems™ Dynabeads™ MAX E. coli O157 Kit
Dynabeads™ MAX E. coli O157 Kit
Brand:  Applied Biosystems™ A10714
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10023393
  • 2136.82 SEK / Each
Estimated Shipment
01-02-2021
Add to basket
For Research Use Only. All usage must comply with product instructions.
Description
Description
Dynabeads™ MAX E.Coli O157 provides a powerful sample preparation method to improve PCR detection and quantification of E.Coli O157:H7 in food, feces, water and other environmental samples.
Based on well known and widely used immunomagnetic separation (IMS) technology, Dynabeads™ MAX E.Coli O157 provides a selective E.Coli O157:H7 capture step that delivers significant improvements in PCR analytical process time, as well as the sensitivity and consistency of results. With Dynabeads™ MAX E.Coli O157, you'll:
  • Perform high throughput, rapid, and effective E.Coli O157:H7 sample preparation
  • Significantly shorten sample pre-enrichment time
  • Improve detection sensitivity in PCR sample analysis
  • Significantly reduce overall analysis time
  • Can be carried out manually or automated using the BeadRetriever™ System
  • Specifications
    Specifications
    Polyclonal
    Bacteria (Intact)
    Immunomagnetic Separation
    Small Volume Microbial Samples
    1-3 mL
    E. coli O157:H7
    Dynabeads® MPC®, KingFisher mL Food Protection Purification System, DynaMag™-2
    Environmental Sample, Feces, Food, Water Samples
    Each unit of product contains 1 ml Dynabeads® MAX anti-E. coli O157 and 16 ml 10X Sample Buffer.
    Store at 2–8°C.
    Not High-throughput Compatible (Manual), Automated Protocols
    50 Tests
    DYNAL®, Dynabeads®
    Room Temperature
    E. coli
    1mL