missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dynamitin (69-376 AA) Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | Dynamitin (69-376 AA) |
---|---|
Concentration | 0.13 mg/mL |
Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16826183
|
Proteintech
10030-2-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16816183
|
Proteintech
10030-2-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This antibody is against the internal region (69-376aa) of full length p50 dynamitin.
This gene encodes a 50-kD subunit of dynactin, a macromolecular complex consisting of 10-11 subunits ranging in size from 22 to 150 kDa. Dynactin binds to both microtubules and cytoplasmic dynein. It is involved in a diverse array of cellular functions, including ER-to-Golgi transport, the centripetal movement of lysosomes and endosomes, spindle formation, chromosome movement, nuclear positioning, and axonogenesis. This subunit is present in 4-5 copies per dynactin molecule. It contains three short alpha-helical coiled-coil domains that may mediate association with self or other dynactin subunits. It may interact directly with the largest subunit (p150) of dynactin and may affix p150 in place.Specifications
Dynamitin (69-376 AA) | |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q13561, Q99KJ8 | |
10540, 69654 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
DCTN 50, DCTN2, DCTN50, dynactin 2, dynactin 2 (p50), Dynactin subunit 2, DYNAMITIN, p50 dynamitin, RBP50 | |
Dctn2 | |
Primary | |
-20°C | |
DCTN2 |