missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Dynorphin A (13-17), Porcine

For cellular and molecular biology applications

Brand:  Alfa Aesar™ J64991.MB

Additional Details : Weight : 0.00025kg

 View more versions of this product

Product Code. 15466679

  • 2622.49 SEK / 25mg


Estimated Shipment date
from Supplier 05-07-2018
Add to basket

Specifications

Specifications

689.72
Porcine
Solid
RUO
Dynorphin A (13-17), porcine
HPLC
25mg
Lys-Trp-Asp-Asn-Gln
-30° to -10°C
Videos
SDS

For Research Use Only